Learn More
Uvex™ 9307 Carbonvision Safety Goggles
19.63 EUR valido fino al 2024-12-31
Usa il codice promo "23255" per avere il tuo prezzo promozionale.
Usa il codice promo "23255" per avere il tuo prezzo promozionale.
Click to view available options
Lens Tint:
Clear
Gray
Materiale del telaio:
Neoprene
Textile
Descrizione
- Frame is constructed with the latest materials and technologies for maximum mechanical stability, while still remaining lightweight
- A TPU layer is firmly welded onto the frame to ensure the perfect fit and allow for easy cleaning
- Certified to EN 166 (personal eye protection) and EN 172 (sunglare filters for industrial use)
- Supravision extreme coating is scratch-proof on the outside, and permanently anti-fog on the inside
- Offers a panoramic view without a tunnel effect due to the close-fitting lens
- Ideal for long periods of wear without pain or pressure points
Specifica
Tipo di lente | Supravision Extreme |
Certifications/Compliance (Europe) | EN 166:2001, EN 172:1994 + A1:2000 + A2:2001 |
Lens Tint | Gray |
Colore telaio | Black, Gray |
Modello | 9307 Carbonvision |
Lens Material | Polycarbonate |
Materiale del telaio | Textile |
UV Protection | UV 400 |
Labeling | W 166 34 B CE - 5-2,5 W 1 B KN CE |