Learn More
Memmert™ Addition of Units for SN75plus
Integration of one additional unit (up to max. 31 units) into an already existent FDA-software licence (only for units with TwinDISPLAY).
Marca: Memmert™ SN75plusFDAQ2
Dettagli aggiuntivi : Peso : 0.01200kg
Descrizione
- Up to max. 31 units
- Only for units with TwinDISPLAY
Specifica
SN75plus Sterilizer |