Learn More
Cole-Parmer™ Filtro in fibra di vetro borosilicato Advantec GC50
Use as pre-filters to extend membrane life, for water/air pollution analysis or liquid clarification. Advantec™ GC50 Borosilicate Glass Fiber Filters are ideal for analytical or gravimetric studies.
Marca: Cole-Parmer™ 06656-03
Dettagli aggiuntivi : Peso : 0.06300kg
Descrizione
Borosilicate glass filters create a biologically inert filter matrix that is resistant to both chemicals and pH extremes. Filters without binder withstand up to 500°C. Filters with binder withstand up to 120°C. Grades GC90, GS25 and DP70 with binder offer higher wet strength and lower fiber release.Specifica
Fiala in vetro borosilicato | |
0.5 μm | |
Confezione da 100 | |
75 mm |
Vetro borosilicato | |
Per l'analisi dell'inquinamento di acqua/aria o per la chiarificazione di liquidi | |
Filtro |