Learn More
PepSlide™ Analyzer One-Year License
Intuitive and easy-to handle software for quantification of microarray data from PEPerCHIP™ Peptide Microarrays.
Marca: PepSlide™ PSA.001.001
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
- The software is adjusted to the PEPperCHIP™ Peptide Microarray platform and links microarray design files to content information for microarray data analysis
- Software supports: quantification of microarray data, data analysis of pre-defined microarray content, microarray autoalignment, state-of-the-art noise and background correction algorithms and spot morphology recognition
Specifica
Software License | |
Quantification of flourescent intensities from PEPperCHIP Peptide Microarrays for antibody validation, epitope mapping, serum screening, biomarker discovery, protein-peptide interaction studies | |
PEPperPRINT™ Peptide Microarrays |