Learn More
BD BBL™ Hemo ID Quad Plate, with Growth Factors
Separates various growth factors by media type. Isolates, differentiates and identifies Haemophilus species based on the X- and V-factor growth requirements and the hemolysis of horse blood.
Marca: BD 297890
Dettagli aggiuntivi : Peso : 0.15000kg
Descrizione
- Facilitates identification and eliminates the difficulties associated with inspecting and interpreting undivided colonies
- After 24 hours of incubation, the Quad Plate will indicate the presence or absence of growth in specific quadrants, depending on X- and V-factor requirements for the various Haemophilus species
Specifica
Hemo ID Quad Plate | |
Growth factors | |
10 Pack |