Learn More
Greiner Bio-One CELLSTAR CELLreactor™ Conical Bottom Polypropylene Filter Top Tube
For shaking suspension cell, aerobic bacteria and microorganisms.
Marca: Greiner Bio-One 227245
Dettagli aggiuntivi : Peso : 0.50000kg
Beschreibung
- Can be used as a small bioreactor for suspension and spheroid cell culture
- Facilitates high number of parallel experiments
- Flexible working volume from 1 to 35mL
- Sterile
- Allows gas exchange
Spezifikation
Culture Tube | |
Free of detectable DNase/RNase, human DNA and pyrogens; non-cytotoxic | |
Polypropylene | |
Shaking suspension cell, aerobic bacteria and microorganisms | |
50 mL | |
Yes |
Blue | |
Filter Screw Cap | |
Yes | |
Sterile | |
Conical Bottom |