Learn More
3M™ Coconut Protein Rapid Kit L25COC
Intended for screening for the presence of coconut proteins in clean-in-place (CIP) final rinse water, environmental swab samples, food ingredients and processed food products.
Marca: 3M™ 7100151334
Dettagli aggiuntivi : Peso : 0.03300kg
Descrizione
- Results visible at 11 ± 1 minute after application of the sample.
- Rapid, qualitative immunochromatographic lateral flow test cartridges designed for the detection of allergenic food proteins.
- Designed to detect both processed and unprocessed allergen proteins.
- Test can accommodate environmental swab, CIP rinse water and food samples.
Specifica
Microbiology Media | |
Allergen | |
25/Cs. |