Learn More
Fisherbrand™ Container with Lid, Side Walls and Closable Holes
For use with HMT 300 MA Autoclaves.
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 2.00000kg
Descrizione
- Solid construction
- 500 x 205 x 185mm
Specifica
Autoclavable | |
HMT 300 MA Autoclaves | |
Autoclave Container with Lid | |
500 x 205 x 185 mm |