Learn More
DARPP-32 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA585787
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Rhesus Monkey (100%), Bovine (100%). Positive Control: mouse brain, rat brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
DARPP-32 is a dopamine (DA) and AMP-regugated ∽32 kDa phosphoprotein that is associated with dopaminoceptive neurons. The protein inhibits Protein Phosphatase I when it is phosphorylated on Thr34. In contrast, when DARPP-32 is phosphorylated on Thr75 the protein acts as an inhibitor of PKA. Phosphorylation of DARPP-32 is thought to play a critical role in the regugation of dopaminergic neurotransmission. In addition, the activity of DARPP-32 is also thought to play important roles in the actions of alcohol, caffeine and Prozac®.Specifica
DARPP-32 | |
Polyclonal | |
Unconjugated | |
Ppp1r1b | |
AU040756, DARPP32, DARPP-32, dopamine- and cAMP-regulated neuronal phosphoprotein, dopamine and cAMP-regulated neuronal phosphoprotein 32, dopamine- and cAMP-regulated phosphoprotein DARPP-32, FLJ20940, IPPD, neuronal phosphoprotein DARPP-32, OTTHUMP00000164275, OTTHUMP00000164276, Ppp1r1b, PPR1B, protein pho, protein phosphatase 1 regulatory inhibitor subunit 1B, protein phosphatase 1 regulatory subunit 1B, protein phosphatase 1, regulatory (inhibitor) subunit 1B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19049, 360616 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot, Western Blot | |
1.11 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300, pH 7 | |
Q60829, Q6J4I0 | |
Ppp1r1b | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of mouse DARPP-32. | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |