Learn More
SCHOTT AG Lighting and Imaging™ Daylight Filter for Halogen Light Sources
The Daylight Filter for Halogen Light Sources Increases the color temperature from a warm halogen light to a cold daylight. For use with focusing lens with 5 mm diameter light guide and KL1500HAL Cold Light Source.
Marca: SCHOTT AG Lighting and Imaging™ 158211
Dettagli aggiuntivi : Peso : 5.00000kg
Descrizione
For use with:
- Focusing lens with 5 mm diameter light guide
- KL1500HAL Cold Light Source
Specifica
Filter | |
Stereo Microscopy Illumination | |
Focusing Lens 158210, KL1500HAL | |
Daylight |