missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHIELD Scientific duoSHIELD™ PFT Nitrile 240
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Descrizione
- Brand signature: Blue for dual risk
- Exam gloves for laboratory general use and for patient protection
- Latex gloves and nitrile gloves
- Non-sterile gloves
- Maximum comfort
- Compliance with the latest European standards
- Category III PPE gloves (PPE Regulation (EU) 2016/425)
- Class 1 MDR (Regulation (EU) 2017/745)
- Length 240 mm minimum
- Biohazard: ISO 374-5:2016
- Biosafety: AQL 0.65 or 1.5 (EN 455-1:2020 and ISO 374-2:2019 Level 2 or 3)
- Virus resistant (ISO 16604:2004 Procedure B)
- Tested for chemical permeation according to ISO 374-1:2016+A1:2018 (Test method EN 16523-1:2015+A1:2018)
- Reduced risk of allergies
- Packaging designed to comply with the requirements of laboratory and medical environments
Specifica
Specifica
Certifications/Compliance (Europe) | PPER (EU) 2016/425, ISO 21420:2020, EN455-2:2015, ISO 374-1:2016-A1:2018 |
Accelerator Free | No |
Applicazioni | Laboratory, Healthcare |
AQL | 0.65 |
Colore | Blue |
Monouso | Single-use |
Finitura esterna | Fingertip Textured |
Finger Thickness (English) | 3.9 mil |
Finger Thickness (Metric) | 0.10 mm |
Latex Free | Yes |
Vedi altri risultati |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'