Learn More
3M™ Edge™ 5 Personal Noise Dosimeter
3M™ Edge™ 5 Personal Noise Dosimeter offers advanced technology packaged in a cable-free, compact frame for a lighter, more ergonomic way to monitor noise levels.
Marca: 3M™ 7000030071
Dettagli aggiuntivi : Peso : 1.81400kg
Descrizione
- Rechargeable lithium polymer battery
- Thin, contoured mount hugs the shoulder; compact frame offers superior comfort
- Intrinsic safety certifications
- Shoulder Mount
Specifica
40 hours | |
88 x 53 x 19 mm | |
Dose, Exposure, Identity Field, L(C-A), Lavg/Leq, Max, Min, Peak, Projected Dose, Run Time, SEL (Lep), SPL, Threshold, TWA, Upper Limit (UL) |
CE, IEC 61252, ANSI S1.25 | |
Docking Station |