Learn More
3M™ Edge Five Bay Docking Station
3M™ Edge Five Bay Docking Station is suitable for use with the Edge Dosimeters.
Marca: 3M™ 7000127801
Dettagli aggiuntivi : Peso : 0.67360kg
Descrizione
- The EdgeDock also downloads data to Detection Management Software (DMS) for reporting, charting and analyzing exposures via the USB port to PC connection
- Multiple EdgeDocks may be interfaced via the USB hub to the PC
- 5-bay EdgeDock docking station for recharge of multiple units
- Quick and easy recharge of Edge dosimeters
- Easy download of data to PC
Specifica
Docking Station Kit | |
(1) 53-575 USB Cable and (1) 17-646 Power Supply |