Learn More
Merck SNAP i.d. 2.0™ IHC Slide Holders
For use with SNAP i.d. 2.0 protein detection system
Marca:  Merck SNAP2SH
Dettagli aggiuntivi : Peso : 0.10000kg
Codice prodotto. 15751917
  • 211.00€ / Pezzo
Spedizione stimata: 28-05-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
  • Disposable slide holders fit standard 25 x 75 x 1mm glass slide
  • Each slide holder features antibody injection/recovery ports to facilitate addition and removal of small volumes of antibodies and reagents
  • Specially designed gaskets eliminate need for pap pens
  • For processing 24 tissue slides
Specifica
Specifica
24/Pk.