Learn More
Fisherbrand™ Electric Bunsen Heating Mantles
Combines advantages of gas burner with clean operation and easy control of electric heating
Marca: Fisherbrand™
179.41 EUR valido fino al 2024-05-31
Usa il codice promo "21638" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.22710kg
Descrizione
The Electric Bunsen combines the advantages of a regular gas burner with the clean easy operation of our Electromantles. Radiation from the heater is directed upwards to a focal point. The Electric Bunsen is ideal for heating test tubes, crucibles, small flasks and beakers, regardless of their shape.
- Burner consumes only 400w of power
- Conical shaped heating element directs radiant heat to top cavity
- Top cowl deflects heat away from your hand
- Air circulation from vented housing keeps base cool enough to hold during operation
- Recommended controller is MC5
- Corrosion-resistant and has cool-to-touch base
Certifications
CE
Servizi
One year
Specifica
CE | |
25 mL | |
115 V | |
Electric Bunsen Burner | |
0.84 oz. |
0.84 oz., 25mL | |
7 in. | |
430w | |
3.5 A |