Learn More
Fisherbrand™ Animal Ear-Punch
Fisherbrand Ear punch are used for punching holes into an animal's ear for labeling purposes.
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 0.01260kg
Descrizione
Fisherbrand Ear punch are used for punching holes into an animal's ear for labeling purposes.
Fisherbrand™ Animal Ear-Punch. These instruments are available in multiple designs, including thumb-style, plier-style, or scissors-style punches. Both the plier-style and scissor-style punches feature a longer handle for improved grip and handling.Ear-Punch
- For use with animal ear tags
- Thumb-type for punching holes into thin part of animal’s ear
- Plier-type: Provides a longer handle for gripping and punching into animal ear
- Made from corrosion-resistant ASTM (899-94) series 400 stainless steel
- Available Ear-punch: Thumb, plier or scissor type
- Compatible with Gamma/ETO/Autoclavable sterilization methods
- Electronically passivated for corrosion resistance
Specifica
Animal Identification | |
Ear Tag Punch | |
2 mm punch hole |