Learn More
Food Calorimeter Replacement Parts
Replacement parts for Food Calorimeter.
Marca: Nickel Electro 16815
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Replacement nickel crucible and igniter coils for Food Calorimeter (Cat. No. S17038).
Specifica
Nickel Crucible | |
Food Calorimeter | |
5.7 cm |
Used for Diluting Alkalis | |
2.24 in. |