Learn More
3M™ Indicatori biologici HEALTH CARE™ Attest
Monitors the effectiveness of the steam sterilization process and provides results within 3 hours. 3M™ Attest™ Rapid Readout Biological Indicators provide the best assurance of sterility by challenging the sterilizer with quantifiable, highly resistant spores.
Marca: 3M™ 7000053661
Dettagli aggiuntivi : Peso : 0.63900kg
Descrizione
- Easy to use and interpret with visual color readout
- Self-contained biological indicators significantly reduce possibility of contamination
- Minimizes false positives
Specifica
Sì | |
EN886-1-2 et ISO 11138-1-3 | |
Indicatore biologico | |
Confezione da 50 |