Learn More
Euro Protection™ Latex Gloves, Powder-Free
Click to view available options
Size:
6
8
10
12
Descrizione
- Liquidproof
- Good dexterity
- Comfort and flexibility
- Suitable for food contact
Specifica
Certifications/Compliance (Europe) | EN374-1:2016K(3/6)TYPE C EN374-5:2016 EN420 |
Monouso | Single-use |
Materiale | Latex |
Powdered | Powder Free |
Size | 12 |
Sterilità | Non-sterile |
Spessore (sistema metrico) | 0.125 mm |
Da utilizzare con (applicazione) | Object handling, food industry, paint, solvent, liquid (non-toxic), assembly, cleaning, hospital environments (laboratory) |
Quantità | 100/Pk |