Learn More
3M™ Molecular Detection Matrix Control MDMC96
Kit contains 96 tests and is used with the 3M Molecular Detection System to check for inhibition of the matrix sample during validation periods, when adopting the 3M method or testing new or unknown matrices.
Marca: 3M™ 7000128756
Dettagli aggiuntivi : Peso : 0.00800kg
Descrizione
- Verify that new or unknown matrices are not inhibitory to amplification.
- Ready-to-use and pre-dispensed reagents.
- Only two transfer steps after enrichment.
- Closed-tube system reduces risk of amplicon contamination.
- Designed to work with 3M proprietary enrichment broths to promote growth.
Specifica
Microbiology Media | |
Environmental Test | |
3 M Molecular Detection System | |
96 |