missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Purified Whole Serum, Sheep
Suitable for use as a serum protein control in immunoelectrophoresis
Marca: MP Biomedicals™ 0855992
104.94 EUR valido fino al 2024-12-31
Usa il codice promo "23257" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.00200kg
Specifica
-20°C | |
Lyophilized | |
2 mL | |
Serum protein control in immunoelectrophoresis, Ouchterlony immunodiffusion, and other immunoprecipitation techniques |
Sheep | |
Sheep Serum | |
60.2mg/mL | |
Yes |