Learn More
Euro Protection™ Nitrile Gloves, Powdered
Catalog Number 15322296
Click to view available options
Size:
7
8
9
10
Euro Protection™ Nitrile Gloves, Powdered provide comfort, flexibility, and dexterity for a variety of tasks including food preparation.
  • Liquidproof
  • Good dexterity
  • Comfort and flexibility
  • Mechanical strength
  • Sweat absorption for powdered version
  • Suitable for food contact
  • Applications: Handling of objects, food industry (preparation), painting, solvent, liquid, assembly of small parts, assembly, cleaning, hospital environments.
Certifications/Compliance (Europe) EN374-1:2016K(6/6)TYPE CEN374-5:2016EN420
Colore Blue
Monouso Single-use
Materiale Nitrile
Powdered Powdered
Product Type Nitrile Examination Glove
Size 7
Sterilità Non-sterile
Spessore (sistema metrico) 0.10 mm
Modello 5906
Quantità 100/Pk.