missing translation for 'onlineSavingsMsg'
Learn More

Merck Novagen™ Lysonase™ Bioprocessing Reagent

Easy, optimized, ready-to-use blend of rLysozyme™ Solution and Benzonase Nuclease. MilliporeSigma™; Lysonase™ Bioprocessing Reagent contains a highly purified and stabilized recombinant lysozyme with specific activity 250 times greater than that of chicken egg white lysozyme.

Marca:  Merck 71230-4

Dettagli aggiuntivi : Peso : 0.20000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 17698685

  • 1285.00€ / Pezzo
Spedizione stimata: 14-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

  • Benzonase Nuclease is a genetically engineered nonspecific endonuclease that degrades all forms of DNA and RNA (single stranded, double stranded, circular, linear)
  • Reduces extract viscosity and increases protein yield
  • Combined activities of rLysozyme and Benzonase Nuclease significantly increase protein extraction efficiency and facilitate downstream processing of protein extracts
  • For efficient protein extraction with BugBuster™ Protein Extraction Reagent, use 10μL Lysonase per 1g cell paste
  • For efficient protein extraction with PopCulture™ Reagent, add 2μL Lysonase per 1mL culture
Specifica

Specifica

Protein Extraction, Protein Preparation
1 mL
Bioprocessing Reagent
-20°C
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali