missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Dicer knockout Mouse Embryonic Stem Cells
Used in common research applications
Marca: Novus Biologicals™ NBP1-96751
Dettagli aggiuntivi : Peso : 0.00970kg
Descrizione
- Cells are supplied in 2mL quantities (about 1 x 106 cells/mL) in freezing media (60% DMEM, 20% FBS, 20% DMSO)
- Gene ID: 23405
- Gene symbol: DICER1
Recommended Storage
Store in gas phase of liquid nitrogen
Servizi
6 Months
This product is for research use only and is not approved for use in humans or in clinical diagnosis