Learn More
Hamilton™ One Hub Assembled
One Hub Assembled with 24 in. of Tubing 22 gauge
Marca: Hamilton™ 8892-01
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Specification Sheet
Fitting Material | CTFE |
---|---|
Tubing Material | PTFE |
Gauge | 22 gauge |
Length | 24 inches |
Sales Quantity | 1 each |
Fitting Type | 1/4 in.-28 |
Number of Hubs | one |
Specifica
1/4 in.-28 in. | |
24 in. | |
Each |