Learn More
PEPperCHIP™ Pan-Corona Spike Protein Microarray
Pan-Corona Spike Protein Microarray with 4,564 specific peptides of seven human coronavirus spike proteins printed in duplicate. The microarray can be used for epitope mapping, serum screening or biomarker discovery.
Marca:  PEPperCHIP™ SPC.052.001
Dettagli aggiuntivi : Peso : 0.02200kg
Codice prodotto. 16638682
  • 560.74€ / Pezzo
Spedizione stimata: 25-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Microarray Content: Spike proteins of SARS-CoV-2, SARS-CoV, MERS-CoV, HCoV-OC43, HCoV-HKU1, HCoV-NL63 and HCoV-229E translated into overlapping peptides
  • Peptide Length/Overlap: 15 aa/ 13 aa
  • Number of Peptides: 4,564 printed in duplicate and HA and polio control peptides
  • Related Products: PEPperCHIP™ Incubation Tray 3/1, PEPperCHIP™ anti-HA control antibodies, MAPIX Analyzer
Specifica
Specifica
Spike Protein Microarray
Epitope Mapping, Serum Screening or Biomarker Discovery
Infectious Disease
Spike Proteins from SARS-CoV-2, SARS-CoV, MERS-CoV, HCoV-OC43, HCoV-HKU1, HCoV-NL63 and HCoV-229 E
1 Slide