Learn More
Lovibond™ Tintometer™ Photometer-System PC MultiDirect
Used in Waste Water, Drinking Water, Industrial Process Water, Scientific Applications, Governmental and Private Laboratories, Universities and Research Centres, Mobile Applications
Marca: Lovibond™ 210000
Dettagli aggiuntivi : Peso : 3.20000kg
Descrizione
- Dual Beam Technology and Interference Filters for highest accuracy
- A wide range of pre-programmed methods
- Long-term stable LEDs as light sources
- Update of new methods and languages via Internet (free of charge)
- Interface
- Memory for 1000 data sets
- Mobile
Specifica
Photometer-System PC Multidirect | |
LED | |
Waste Water, Drinking Water, Industrial Process Water, Scientific Applications, Governmental and Private Laboratories, Universities and Research Centres, Mobile Applications |