Learn More
Quickfit™ Microscale Kit
Designed for organic chemistry experiments using milligram quantities of reagents
Marca: Quickfit ML1
261.25 EUR valido fino al 2024-12-31
Usa il codice promo "23259" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 1.17000kg
Descrizione
- Comprises 12 items of glassware that all feature 14/10 joint sizes for compatibility
- Utilise combination of standard Quickfit™ ground glass joints together with screwthread couplings, sealing 'O' rings and plastic screw caps
- Screwthread connector system eliminates requirement for grease, saves on space, using clamps and vastly reduces danger of glassware becoming loose during reactions
- Allows construction of many assemblies including reflux, distillation, moisture sensitive reactions and fractional distillation
- Housed in sturdy Gratnells tray with protective foam insert
Specifica
Microscale Kit | |
Air condenser, Jacketed condenser, Drying tube, Still head, Thermometer adapter, Receiver, Erlenmeyer flask 10mL, Erlenmeyer flask 25mL, Round bottom flask 10mL, Round bottom flask 25mL, Pear shaped flask 10mL, Pear shaped flask 25mL, Bag of spare caps, 'O' rings, sealing discs |