Learn More
Quickfit™ Organic Chemistry Set
Starter kit - ideal for introducing students to principal techniques of preparative organic chemistry
Marca:  Quickfit 29BU/M
159.60 EUR valido fino al 2024-12-31
Usa il codice promo "23259" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.48000kg
Codice prodotto. 12103112
  • 168.00€ / Pezzo
Spedizione stimata: 01-07-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Starter kit - ideal for introducing students to principal techniques of preparative organic chemistry
  • Comprises 5 glassware items all with size 14/23 Quickfit™ ground glass joints - giving maximum flexibility
  • Scale based on starting material quantities up to 30g
  • Both reflux and distillation assemblies can be constructed
  • Housed in a robust cardboard box with foam rubber liner for product protection
Specifica
Specifica
Organic chemistry kit
Five items of Glassware includes 50mL pear shaped flask, two still heads, three Liebig condenser, four screw cap adapters, five receiver adapter
Ideal for introducing students to the main techniques of preparative organic chemistry