Learn More
Euro Protection™ Revlux Welder Goggles
Click to view available options
Green Frame
Descrizione
- PVC Frame
- Lens support in black ABS
- Indirect ventilation
- Adjustable headband
- Protane 5 lens
- Clear and IR5 lenses
- Frame marking 3 4 ft.
- Lens marking 2 - 1.2 ft.
Specifica
Certificazione/Conformità | EN 166, EN 169, EN 170 |
Lens Tint | IR/PC 5.0 |
Da utilizzare con (applicazione) | Welding |
Colore telaio | Green |
Include | Clear and IR5 lenses |
Materiale del telaio | PVC |