missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARS-CoV-2 Spike Protein (RBD) (aa330-524), His Tag Recombinant Protein
HEK293 cells Recombinant Protein
Marca: Invitrogen™ RP87687
2956.10 EUR valido fino al 2024-06-28
Usa il codice promo "21649" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold symptoms to severe effects including death and respiratory distress. The CoV Spike (S) protein plays the most important roles in viral attachment, fusion and entry, and serves as a target for development of antibodies, entry inhibitors and vaccines. Of note is the Spike protein receptor-binding domain (RBD, S-RBD) in SARS-CoV-2 Spike protein which binds strongly to human and bat angiotensin-converting enzyme 2 (ACE2) receptors. SARS-CoV-2 RBD exhibits significantly higher binding affinity to ACE2 receptor than SARS-CoV RBD in comparison. S1 contains a receptor-binding domain (RBD) that can specifically bind to angiotensin-converting enzyme 2 (ACE2), the receptor on target cells.Specifica
Control, ELISA, Western Blot | |
SARS-CoV-2 Spike Protein (S-RBD) (aa330-524), His Tag | |
HEK293 cells | |
RUO | |
Virus | |
Recombinant | |
>95% |
PBS with no preservative | |
1 mg | |
-20°Cor to +80°C if preferred | |
Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2 | |
Unconjugated | |
Lyophilized |