missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHIELD Scientific SHIELDskin™ ORANGE NITRILE™ 300
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Descrizione
- Brand signature: Orange for biological risk
- Laboratory gloves for biohazard protection
- Latex gloves and nitrile/neoprene gloves
- Non-sterile and sterile gloves
- Maximum comfort
- Compliance with the latest European standards
- Category III PPE gloves (PPE Regulation (EU) 2016/425)
- Extra length from 260 to 300 mm (meets or exceeds ISO 21420:2020+A1:2022)
- Biohazard: ISO 374-5:2016
- Biosafety: AQL 0.25 (ISO 374-2:2019 Level 3)
- Virus resistant (ISO 16604:2004 Procedure B)
- Extensively tested for chemical permeation according to ISO 374-1:2016+A1:2018 (Test method EN 16523-1:2015+A1:2018)
- Reduced risk of allergies
- Double-walled protection afforded by the twinSHIELD™ technology
- Packaging designed to comply with laboratory environments processes
Specifica
Specifica
Certificazione/Conformità | AQL 0.25 |
Certifications/Compliance (Europe) | PPER (EU) 2016/425, ISO 21420:2020, EN455-2:2015, ISO 374-1:2016-A1:2018 |
Accelerator Free | Yes |
Applicazioni | Laboratory |
AQL | 0.25 |
Colore | Orange |
Monouso | Single-use |
Finitura esterna | Fingertip Textured |
Finger Thickness (English) | 6.7 mil |
Finger Thickness (Metric) | 0.17 mm |
Vedi altri risultati |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'