missing translation for 'onlineSavingsMsg'
Learn More

SK-BR-3 (human breast adenocarcinoma) Nuclear extract lysate, Denatured; Abnova

Marca:  Abnova L050V3.50ug

Dettagli aggiuntivi : Peso : 0.02000kg

Codice prodotto. 16089625

  • 209.00€ / 50µg
Spedizione stimata: 24-06-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

  • Tissue: Breast
  • Host: Human
  • Lysis buffer: Buffer A (10mM HEPES pH 7.9, 1.5mM MgCl2, 10mM KCl, 0.5 mM DTT), Buffer C (20mM HEPES pH 7.9, 25%(v/v) Glycerol , 0.42M NaCl , 1.5mM MgCl2, 0.2mM EDTA, 0.5mM DTT and 0.5mM PMSF), Buffer D (20mM HEPES pH 7.9, 20%(v/v) glycerol, 50mM KCl, 0.2mM EDTA, 0.5mM DTT and 0.5mM PMSF)
  • Storage buffer: In ready-to-use 1X sample buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% Bromophenol blue)

Applications:

Western Blotting

Specifica

Specifica

Western Blot
Nuclear extract cell lysate (denatured)
Nuclear extract was prepared by using a modified protocol of Dignam et al. Cells were Harvested and homogenized in Buffer A, and then centrifugated at 25,000 g for 20 minutes to remove cytoplasm and pellet the nuclei. The pellet was re-suspended in Buffer C, and then the suspensions were centrifuged to collect nuclear extract. The supernatant was dialyzed against Buffer D. The dialysate was then centrifuged, divided into aliquots, and stored at -80°C. The protein concentration was determined by the method of Bradford (Bio-Rad protein assay, microplate standard assay). The lysate was adjusted to 2.5 mg/ml, and then mixed with 5X Sample Buffer to become final 2 mg/ml in 1X Sample Buffer. The lysate was heated at 95°C for 5 min, and cooled rapidly.
50 μg
Human
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Breast
Buffer A: 10mM HEPES pH 7.9, 1.5mM MgCl2, 10mM KCl, 0.5 mM DTT. Buffer C: 20mM HEPES pH 7.9, 25%(v/v) Glycerol , 0.42M NaCl , 1.5mM MgCl2, 0.2 mM EDTA, 0.5 mM DTT & 0.5 mM PMSF. Buffer D : 20mM HEPES pH 7.9, 20%(v/v) glycerol, 50mM KCl, 0.2 mM EDTA, 0.5 mM DTT & 0.5 mM PMSF.
12.5% SDS-PAGE Stained with Coomassie Blue.
In ready-to-use 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue).
2 mg/mL
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali