Learn More
Fisherbrand™ Lame per rasoi di sicurezza
Say no to dangerous sharps. SmartSlicer™ Safety Razor Blades won't scratch surfaces when cutting agarose gels.
Marca: Fisherbrand™
28.12 EUR valido fino al 2024-06-30
Usa il codice promo "21742" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.05760kg
Descrizione
- Cleanly slice agarose gels* right on the light box without scratching the surface.
- Ideal for cutting soft tissue and plant matter.
- Adjust the size required by simply cutting with scissors.
- Available in two sizes and made in the USA from food grade PET.
- Supplied in packs of 100.
Specifica
PET per alimenti | |
Taglio gel di agarosio, taglio di tessuti molli e vegetali | |
Sì | |
0,22 mm | |
100/conf. |
Blade | |
Small | |
Non-sterile | |
Food Grade | |
No |