Learn More
Thermo Scientific™ Standard Fixed Needle Syringes for Rheodyne/Valco™ Injectors
Thermo Scientific™ Standard Fixed Needle Syringes for Rheodyne/Valco Injectors offer easy, accurate and reproducible manual injections.
Marca:  Thermo Scientific™ 365GL251
Dettagli aggiuntivi : Peso : 0.10000kg
Codice prodotto. 10171073
  • 60.95€ / Pezzo
Spedizione stimata: 24-06-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
Thermo Scientific™ Standard Fixed Needle Syringes for Rheodyne/Valco Injectors offer easy, accurate and reproducible manual injections.
  • Square tip to prevent damage to the injector
  • Wide range of volumes
  • Precision made from borosilicate glass and stainless steel
  • Robust design and easy-to-read markings
Specifica
Specifica
50uL
50 mm
No
90° Blunt
22 G
HPLC Autosampler Syringe
50 μL
Injector