Learn More
Corning™ Tank Liners
Designed to fit standardized cylindrical tanks manufactured by Nalge™/Nunc™ and other brands
Marca: Corning™ 91-300-60
Dettagli aggiuntivi : Peso : 1.29390kg
Descrizione
- Made from low-density polyethylene (LDPE)
- Eliminate costs associated with cleaning and validation of tanks
- Decrease labor costs and increase turnaround time
- Eliminate risks associated with cross contamination
- Help extend life of reuseable tanks, resulting in reduced capital expenditures
- Gamma irradiated, individually packaged
- Choice of gusseted and non-gusseted available
- Available in many sizes
Specifica
Non-Gusseted Tank Liner | |
560 L | |
LDPE |