missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Telenzepine dihydrochloride, Tocris Bioscience™
Click to view available options
Quantità:
50 mg
Descrizione
Selective high affinity muscarinic M1 receptor antagonist (Ki = 0.94 nM). Inhibits gastric acid secretion.
Specifica
Materiale o nome chimico | Telenzepine dihydrochloride |
Quantità | 50 mg |
Peso formulazione | 443.39 |
Percent Purity | >98% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'