Learn More
WTW™ Cella conduttimetrica a 4 elettrodi TetraCon™ 925
With its easy handling and robust design, these electrode conductivity cells are proven 2 and 4 electrode technology. The four electrode measuring cell TetraCon™ 925 model is for medium and high conductivity.
Marca: WTW™ 301710
514.21 EUR valido fino al 2024-05-31
Usa il codice promo "21700" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.22500kg
Descrizione
- Graphite four electrode measuring cell, 1.5 fixed cable
- IDS conductivity measuring cells – measurements from pure water all the way to concentrated solutions with proven two and four electrode technology
- Easiest handling, robust design
- Wide area of application - from pure water to highly concentrated solutions
- The four electrode measuring cell TetraCon™ 925 model is for medium and high conductivity
Specifica
4-electrodes, Graphite | |
da 0 a 100 °C | |
120 mm |
1.5 m | |
15.3 mm |