missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human FADD siRNA
Blocks transcription and translation of FADD from Human.
250.00€ - 369.00€
Specifica
Numero di accesso | NP_003815 |
---|---|
Cell Lines Tested | 293 Human Embryonic Kidney Cells with NF-kappa B and Flag tagged DR4 constructs. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Simbolo del gene | FADD |
ID gene (immissione) | 8772 |
Specifica
NP_003815 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
8772 | |
Fas Associated Via Death Domain, Fas-Associating Death Domain-Containing Protein, Growth-Inhibiting Gene 3 Protein, Fas (TNFRSF6)-Associated via Death Domain, Protein FADD, MORT1, FAS-Associated Death Domain Protein | |
Lyophilized |
293 Human Embryonic Kidney Cells with NF-kappa B and Flag tagged DR4 constructs. | |
FADD | |
siRNA | |
Human |
Sicurezza e movimentazione
missing translation for 'shelfLife' : This product is guaranteed to work for a period of two years when stored at -70°C or colder, and one year when aliquoted and stored at -20°C.