Learn More
Velp Scientifica™ OXISoft™ Software
Velp Scientifica™ OXISoft™ Software manages all analysis steps and readily provides valuable information to scientists, for use with OHS Controllers
Marca: Velp Scientifica™ A00000391
Dettagli aggiuntivi : Peso : 0.50000kg
Descrizione
- This software manages all analysis steps and readily provides valuable information to scientists conducting experiments
- Enables scientists to perform repeatability tests, freshness tests, formula comparison, packaging comparison, IP during ageing, and estimated shelf life
- Up to four tests can be managed by the same software for maximum productivity
Specifica
OHS Controllers | |
Software |