CAS RN 1000520-92-2
IUPAC Nome:
2-(4-fluoro-3-methylphenyl)acetic acid
Sinonimi:
(4-fluoro-3-methylphenyl)acetic acid
Peso molecolare (g/mol):
168.17
Formula molecolare:
C9H9FO2
InChi Key:
VZDGZGNZGMJNPR-UHFFFAOYSA-N
SMILES:
CC1=C(F)C=CC(CC(O)=O)=C1
1
–
1
di
1
risultati
1
4-Fluoro-3-methylphenylacetic acid, 97%, Thermo Scientific™
CAS: 1000520-92-2 Formula molecolare: C9H9FO2 Molecular Weight (g/mol): 168.167 Numero MDL: MFCD09832241 InChI Key: VZDGZGNZGMJNPR-UHFFFAOYSA-N Sinonimo: 2-4-fluoro-3-methylphenyl acetic acid,4-fluoro-3-methylphenylacetic acid,4-fluoro-3-methylphenyl acetic acid,4-fluoro-3-methylphenylacetic aicd PubChem CID: 39103878 IUPAC Name: 2-(4-fluoro-3-methylphenyl)acetic acid SMILES: CC1=C(C=CC(=C1)CC(=O)O)F