CAS RN 10022-13-6
IUPAC Nome:
[3,4,6-tris(acetyloxy)-5-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)oxan-2-yl]methyl acetate
Sinonimi:
[3,4,6-tris(acetyloxy)-5-(1,3-dioxoisoindol-2-yl)oxan-2-yl]methyl acetate
Peso molecolare (g/mol):
477.42
Formula molecolare:
C22H23NO11
InChi Key:
DUXJAHFLYZUOPT-UHFFFAOYNA-N
SMILES:
CC(=O)OCC1OC(OC(C)=O)C(C(OC(C)=O)C1OC(C)=O)N1C(=O)C2=CC=CC=C2C1=O
1
–
3
di
3
risultati
1
2-Deoxy-2-N-phthalimido-1,3,4,6-tetra-O-acetyl-Beta-D-glucopyranose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Deoxy-2-N-phthalimido-1,3,4,6-tetra-O-acetyl-Beta-D-glucopyranose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Deoxy-2-N-phthalimido-1,3,4,6-tetra-O-acetyl-Beta-D-glucopyranose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.