CAS RN 1007882-23-6
CAS RN 1007882-23-6
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
(2S,2'S)-2,2'-([1,1'-Biphenyl]-4,4'-diyldi-1H-imidazole-5,2-diyl)bis-1-pyrrolidinecarboxylic Acid 1,1'-Bis(1,1-dimethylethyl) Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(2S,2'S)-2,2'-([1,1'-Biphenyl]-4,4'-diyldi-1H-imidazole-5,2-diyl)bis-1-pyrrolidinecarboxylic Acid 1,1'-Bis(1,1-dimethylethyl) Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(2S,2'S)-2,2'-([1,1'-Biphenyl]-4,4'-diyldi-1H-imidazole-5,2-diyl)bis-1-pyrrolidinecarboxylic Acid 1,1'-Bis(1,1-dimethylethyl) Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.