CAS RN 1009093-14-4
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
2
di
2
risultati
1
N-[(1R,2R)-2-Hydroxy-1-(hydroxymethyl)-2-phenylethyl]carbamic Acid tert-Butyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-[(1R,2R)-2-Hydroxy-1-(hydroxymethyl)-2-phenylethyl]carbamic Acid tert-Butyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.