CAS RN 101-76-8
CAS RN 101-76-8
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
4,4'-DDM, TRC
CAS: 101-76-8 Formula molecolare: C13 H10 Cl2 Molecular Weight (g/mol): 237.12 Sinonimo: Methane, bis(p-chlorophenyl)- (6CI,7CI,8CI),1,1'-Methylenebis[4-chlorobenzene],4,4'-Dichlorodiphenylmethane,Bis(4-chlorophenyl)methane,Bis(p-chlorophenyl)methane,DBM,DDM,NSC 406594,p,p'-DDM,p,p'-Dichlorodiphenylmethane IUPAC Name: 1-chloro-4-[(4-chlorophenyl)methyl]benzene SMILES: Clc1ccc(Cc2ccc(Cl)cc2)cc1