CAS RN 10351-76-5
CAS RN 10351-76-5
IUPAC Name:
6-methyl-1H-1,3-benzodiazole-5-carboxylic acid hydrochloride
Sinonimi:
6-methyl-1H-1,3-benzodiazole-5-carboxylic acid hydrochloride
Peso molecolare (g/mol):
212.63
Formula molecolare:
C9H9ClN2O2
InChi Key:
QCJPDBNTTAJIRH-UHFFFAOYSA-N
SMILES:
Cl.CC1=C(C=C2N=CNC2=C1)C(O)=O
1
–
1
di
1
risultati
1
6-Methylbenzimidazole-5-carboxylic acid hydrochloride, 97%, Thermo Scientific Chemicals
CAS: 10351-76-5 Formula molecolare: C9H9ClN2O2 Molecular Weight (g/mol): 212.633 Numero MDL: MFCD09909565 InChI Key: QCJPDBNTTAJIRH-UHFFFAOYSA-N Sinonimo: 6-methyl-1h-1,3-benzodiazole-5-carboxylic acid,6-methyl-5-benzimidazole carboxylic acid,1h-benzimidazole-5-carboxylic acid,6-methyl,1h-benzimidazole-5-carboxylicacid, 6-methyl,1h-benzimidazole-6-carboxylicacid, 5-methyl,5-methyl-3h-benzoimidazole-6-carboxylic acid,5-methyl-1h-benzo d imidazole-6-carboxylic acid,1h-benzimidazole-5-carboxylic acid, 6-methyl-9ci PubChem CID: 45925698 IUPAC Name: 6-methyl-1H-benzimidazole-5-carboxylic acid;hydrochloride SMILES: CC1=CC2=C(C=C1C(=O)O)N=CN2.Cl