CAS RN 105494-69-7
IUPAC Nome:
1-methyl-2-(tributylstannyl)-1H-imidazole
Sinonimi:
1-methyl-2-(tributylstannyl)imidazole
Peso molecolare (g/mol):
371.16
Formula molecolare:
C16H32N2Sn
InChi Key:
KFWFYOPKNSYTMV-UHFFFAOYSA-N
SMILES:
CCCC[Sn](CCCC)(CCCC)C1=NC=CN1C
1
–
1
di
1
risultati
1
1-Methyl-2-(tri-n-butylstannyl)imidazole, 90+%, Thermo Scientific Chemicals
CAS: 105494-69-7 Formula molecolare: C16H32N2Sn Molecular Weight (g/mol): 371.156 Numero MDL: MFCD01319030 InChI Key: KFWFYOPKNSYTMV-UHFFFAOYSA-N Sinonimo: 1-methyl-2-tributylstannyl-1h-imidazole,1-methyl-2-tributylstannyl imidazole,1-methyl-2-tri-n-butylstannyl imidazole,1-methyl-1h-imidazol-2-yl tributylstannane,1-methyl-2-tributylstannanyl-1h-imidazole,acmc-20aotl,tributyl-1-methylimidazol-2-yl stannane,n-methyl 2-tributylstannylimidazole,2-tri-n-butylstannyl-n-methylimidazole PubChem CID: 2763252 IUPAC Name: tributyl-(1-methylimidazol-2-yl)stannane SMILES: CCCC[Sn](CCCC)(CCCC)C1=NC=CN1C