CAS RN 107-50-6
IUPAC Nome:
tetradecamethylcycloheptasiloxane
Sinonimi:
tetradecamethylcycloheptasiloxane
Peso molecolare (g/mol):
519.08
Formula molecolare:
C14H42O7Si7
InChi Key:
GSANOGQCVHBHIF-UHFFFAOYSA-N
SMILES:
C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O1
1
–
2
di
2
risultati
1
Tetradecamethylcycloheptasiloxane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Tetradecamethylcycloheptasiloxane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.