CAS RN 1135389-59-1
IUPAC Nome:
(1,3-dioxolan-2-yl)methanide; bromozincylium
Sinonimi:
1,3-dioxolan-2-ylmethanide; bromozincylium
Peso molecolare (g/mol):
232.38
Formula molecolare:
C4H7BrO2Zn
InChi Key:
ZEBRGOCCZOTNDU-UHFFFAOYSA-M
SMILES:
[Zn+]Br.[CH2-]C1OCCO1