CAS RN 1171123-46-8
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
2
di
2
risultati
1
R&D Systems™ EMD 386088 hydrochloride
A Tocris product, part of R&D Systems. Potent 5-HT6 agonist
2